Assay Details
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012670.2 | NP_001012688.1 | 9 - 10 | 1898 | 72 | |
GenBank mRNA | AB072368.1 | - | 9 - 10 | 1865 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
