Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013592.1 | NP_001013610.1 | 6 - 7 | 682 | 82 | |
GenBank mRNA | BC104579.1 | - | 5 - 6 | 669 | 82 | |
BT021005.1 | - | 6 - 7 | 682 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
