Assay Details
Gene Symbol: RHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014890.2 | NP_001014890.1 | 2 - 3 | 531 | 92 | |
GenBank mRNA | M21606.1 | - | 2 - 3 | 516 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
