Assay Details
Gene Symbol: PNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014907.1 | NP_001014907.1 | 5 - 6 | 661 | 141 | |
GenBank mRNA | BC103208.1 | - | 5 - 6 | 676 | 141 | |
BT020917.1 | - | 5 - 6 | 661 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
