Assay Details
Gene Symbol: SSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015533.1 | NP_001015533.1 | 4 - 5 | 425 | 94 | |
GenBank mRNA | BC102097.1 | - | 4 - 5 | 407 | 94 | |
BC151423.1 | - | 4 - 5 | 406 | 94 | ||
BT020976.1 | - | 4 - 5 | 425 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
