Assay Details
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015548.2 | NP_001015548.1 | 1 - 2 | 95 | 51 | |
GenBank mRNA | AY911358.1 | - | 1 - 2 | 50 | 51 | |
BC102493.1 | - | 1 | 50 | 51 | ||
BT021025.1 | - | 1 - 2 | 37 | 51 | ||
EE223032.1 | - | 1 - 2 | 96 | 51 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102493.1, NM_001015548.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
