Assay Details
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015548.2 | NP_001015548.1 | 3 - 4 | 432 | 91 | |
GenBank mRNA | AY911358.1 | - | 3 - 4 | 387 | 91 | |
BC102493.1 | - | 1 | 387 | 91 | ||
BT021025.1 | - | 3 - 4 | 374 | 91 | ||
EE223032.1 | - | 3 - 4 | 433 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
