Assay Details
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015548.2 | NP_001015548.1 | 5 - 6 | 741 | 76 | |
GenBank mRNA | AY911358.1 | - | 5 - 6 | 696 | 76 | |
BC102493.1 | - | 1 | 696 | 76 | ||
BT021025.1 | - | 5 - 6 | 683 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
