Assay Details
Gene Symbol: TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015554.1 | NP_001015554.1 | 3 - 4 | 415 | 60 | |
GenBank mRNA | BC140528.1 | - | 3 - 4 | 425 | 60 | |
BT020758.1 | - | 3 - 4 | 415 | 60 | ||
BT021559.1 | - | 3 - 4 | 545 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
