Assay Details
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015555.1 | NP_001015555.1 | 11 - 12 | 1317 | 79 | |
GenBank mRNA | BC102888.1 | - | 11 - 12 | 1263 | 79 | |
BT021065.1 | - | 11 - 12 | 1317 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
