Assay Details
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025329.2 | NP_001020500.1 | 5 - 6 | 503 | 69 | |
GenBank mRNA | AY911362.1 | - | 5 - 6 | 504 | 69 | |
BC109650.1 | - | 5 - 6 | 562 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
