Assay Details
Gene Symbol: C17H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031753.1 | NP_001026923.1 | 4 - 5 | 373 | 124 | |
GenBank mRNA | BT021875.1 | - | 4 - 5 | 373 | 124 | |
BT021897.1 | - | 4 - 5 | 373 | 124 | ||
BT029879.1 | - | 4 - 5 | 362 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
