Assay Details
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034254.2 | NP_001029426.1 | 5 - 6 | 462 | 128 | |
GenBank mRNA | BC103163.1 | - | 5 - 6 | 463 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
