Assay Details
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034285.2 | NP_001029457.1 | 5 - 6 | 475 | 101 | |
GenBank mRNA | BC102636.1 | - | 5 - 6 | 476 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
