Assay Details
Gene Symbol: CNOT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034312.1 | NP_001029484.1 | 5 - 6 | 895 | 135 | |
GenBank mRNA | BC103000.1 | - | 5 - 6 | 895 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
