Assay Details
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034405.2 | NP_001029577.1 | 5 - 6 | 1454 | 102 | |
GenBank mRNA | BC103441.1 | - | 5 - 6 | 1455 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
