Assay Details
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034478.1 | NP_001029650.1 | 1 - 2 | 106 | 108 | |
GenBank mRNA | BC103304.1 | - | 1 - 2 | 106 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103304.1, NM_001034478.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
