Assay Details
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034507.2 | NP_001029679.1 | 4 - 5 | 498 | 93 | |
GenBank mRNA | BC102506.1 | - | 4 - 5 | 499 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
