Assay Details
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034644.2 | NP_001029816.1 | 3 - 4 | 534 | 148 | |
GenBank mRNA | BC102111.1 | - | 3 - 4 | 534 | 148 | |
BT025381.1 | - | 3 - 4 | 586 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
