Assay Details
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034740.2 | NP_001029912.1 | 3 - 4 | 294 | 107 | |
GenBank mRNA | BC102499.1 | - | 3 - 4 | 295 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
