Assay Details
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034780.1 | NP_001029952.1 | 3 - 4 | 362 | 72 | |
GenBank mRNA | BT020923.1 | - | 3 - 4 | 362 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
