Assay Details
Gene Symbol: PDGFRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035301.1 | NP_001030378.1 | 5 - 6 | 1056 | 100 | |
GenBank mRNA | BC113220.1 | - | 5 - 6 | 1004 | 100 | |
BT021182.1 | - | 5 - 6 | 1056 | 100 | ||
BT026299.1 | - | 5 - 6 | 923 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
