Assay Details
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037142.1 | NP_001032219.1 | 2 - 3 | 229 | 75 | |
GenBank mRNA | BC151327.1 | - | 3 - 4 | 423 | 75 | |
BT020888.1 | - | 2 - 3 | 229 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
