Assay Details
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174357.3 | NP_776782.1 | 3 - 4 | 339 | 127 | |
GenBank mRNA | AB005148.1 | - | 3 - 4 | 341 | 127 | |
BC108128.1 | - | 2 - 3 | 2001 | 127 | ||
BC134577.1 | - | 4 - 5 | 320 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
