Assay Details
Gene Symbol: KRT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174377.2 | NP_776802.2 | 4 - 5 | 974 | 76 | |
GenBank mRNA | BC149077.1 | - | 4 - 5 | 978 | 76 | |
X01460.1 | - | 134 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
