Assay Details
Gene Symbol: VEGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174487.2 | NP_776912.1 | 4 - 5 | 465 | 68 | |
GenBank mRNA | AB004273.1 | - | 4 - 5 | 465 | 68 | |
AB004274.1 | - | 4 - 5 | 330 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
