Assay Details
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174514.2 | NP_776939.1 | 2 - 3 | 449 | 84 | |
GenBank mRNA | AF487464.1 | - | 2 - 3 | 449 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
