Assay Details
Gene Symbol: IGFBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174556.1 | NP_776981.1 | 4 - 5 | 1027 | 77 | |
GenBank mRNA | AF305199.1 | - | 4 - 5 | 1008 | 77 | |
BC149336.1 | - | 4 - 5 | 1028 | 77 | ||
M76478.1 | - | 4 - 5 | 1010 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
