Assay Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180996.1 | NP_851339.1 | 4 - 5 | 512 | 95 | |
GenBank mRNA | AY912488.1 | - | 3 - 4 | 372 | 95 | |
DQ184480.1 | - | 3 - 4 | 460 | 95 | ||
EF451794.1 | - | 3 - 4 | 450 | 95 | ||
EF470979.1 | - | 3 - 4 | 369 | 95 | ||
FN994193.1 | - | 3 - 4 | 360 | 95 | ||
KM386997.1 | - | 3 - 4 | 372 | 95 | ||
KP221576.1 | - | 4 - 5 | 477 | 95 | ||
M23813.1 | - | 4 - 5 | 95 | |||
M27325.1 | - | 4 - 5 | 477 | 95 | ||
MK309337.1 | - | 1 - 2 | 173 | 95 | ||
V00111.1 | - | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
