Assay Details
Gene Symbol: MRPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201606.2 | NP_963900.1 | 2 - 3 | 293 | 78 | |
GenBank mRNA | AB116379.1 | - | 2 - 3 | 308 | 78 | |
BC134767.1 | - | 2 - 3 | 317 | 78 | ||
BC142005.1 | - | 2 - 3 | 317 | 78 | ||
BT020735.1 | - | 2 - 3 | 270 | 78 | ||
BT020755.1 | - | 2 - 3 | 305 | 78 | ||
BT020982.1 | - | 2 - 3 | 293 | 78 | ||
BT030597.1 | - | 2 - 3 | 364 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
