Assay Details
Gene Symbol: MRPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201606.2 | NP_963900.1 | 4 - 5 | 509 | 82 | |
GenBank mRNA | AB116379.1 | - | 4 - 5 | 524 | 82 | |
BC134767.1 | - | 4 - 5 | 533 | 82 | ||
BC142005.1 | - | 4 - 5 | 533 | 82 | ||
BT020735.1 | - | 4 - 5 | 486 | 82 | ||
BT020755.1 | - | 4 - 5 | 521 | 82 | ||
BT020982.1 | - | 4 - 5 | 509 | 82 | ||
BT030597.1 | - | 4 - 5 | 580 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
