Assay Details
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025334.2 | NP_001020505.1 | 1 - 2 | 122 | 96 | |
GenBank mRNA | AY911367.1 | - | 1 - 2 | 94 | 96 | |
BC102283.1 | - | 1 - 2 | 122 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
