Assay Details
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025334.2 | NP_001020505.1 | 3 - 4 | 368 | 64 | |
GenBank mRNA | AY911367.1 | - | 3 - 4 | 340 | 64 | |
BC102283.1 | - | 3 - 4 | 368 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
