Assay Details
Gene Symbol: ZNF148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033121.1 | NP_001028293.1 | 1 - 2 | 80 | 117 | |
GenBank mRNA | DQ162621.1 | - | 1 - 2 | 80 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001033121.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
