Assay Details
Gene Symbol: RPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033613.2 | NP_001028785.1 | 5 - 6 | 576 | 96 | |
GenBank mRNA | AB098951.1 | - | 4 - 5 | 485 | 96 | |
AF013215.1 | - | 4 - 5 | 532 | 96 | ||
BC102227.1 | - | 5 - 6 | 578 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
