Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034040.2 | NP_001029212.1 | 6 - 7 | 658 | 76 | |
GenBank mRNA | BC102272.1 | - | 6 - 7 | 661 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
