Assay Details
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034042.2 | NP_001029214.1 | 3 - 4 | 301 | 107 | |
GenBank mRNA | AB098920.1 | - | 2 - 3 | 285 | 107 | |
AB098925.1 | - | 2 - 3 | 286 | 107 | ||
BC103159.1 | - | 3 - 4 | 302 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
