Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034247.1 | NP_001029419.1 | 1 - 2 | 247 | 103 | |
GenBank mRNA | BC103198.1 | - | 1 - 2 | 273 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103198.1, NM_001034247.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
