Assay Details
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034271.2 | NP_001029443.1 | 4 - 5 | 296 | 103 | |
GenBank mRNA | BC102590.1 | - | 4 - 5 | 297 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
