Assay Details
Gene Symbol: MRPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034343.2 | NP_001029515.1 | 1 - 2 | 113 | 79 | |
GenBank mRNA | BC102580.1 | - | 1 - 2 | 114 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
