Assay Details
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034388.2 | NP_001029560.1 | 1 - 2 | 95 | 56 | |
GenBank mRNA | BC102963.1 | - | 1 - 2 | 96 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
