Assay Details
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034407.1 | NP_001029579.1 | 10 - 11 | 1406 | 100 | |
GenBank mRNA | BC103405.1 | - | 10 - 11 | 1406 | 100 | |
BT030568.1 | - | 10 - 11 | 1390 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
