Assay Details
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034407.1 | NP_001029579.1 | 5 - 6 | 823 | 74 | |
GenBank mRNA | BC103405.1 | - | 5 - 6 | 823 | 74 | |
BT030568.1 | - | 5 - 6 | 806 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
