Assay Details
Gene Symbol: CXCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034410.2 | NP_001029582.1 | 3 - 4 | 474 | 78 | |
GenBank mRNA | BC103335.1 | - | 3 - 4 | 475 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
