Assay Details
Gene Symbol: RPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034427.2 | NP_001029599.1 | 1 - 2 | 114 | 112 | |
GenBank mRNA | BC103268.1 | - | 1 - 2 | 115 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
