Assay Details
Gene Symbol: DCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034573.1 | NP_001029745.1 | 1 - 2 | 240 | 95 | |
GenBank mRNA | BC105141.1 | - | 1 - 2 | 240 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
