Assay Details
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034591.2 | NP_001029763.1 | 1 - 2 | 426 | 69 | |
GenBank mRNA | BC102859.1 | - | 1 - 2 | 427 | 69 | |
BT030542.1 | - | 1 - 2 | 324 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
