Assay Details
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034605.2 | NP_001029777.1 | 1 | 352 | 76 | |
GenBank mRNA | BC102692.1 | - | 1 | 354 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
