Assay Details
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034625.2 | NP_001029797.1 | 1 | 534 | 76 | |
GenBank mRNA | BC102277.1 | - | 1 | 535 | 76 | |
BC151407.1 | - | 4 - 5 | 536 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
