Assay Details
Gene Symbol: MRPL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034641.2 | NP_001029813.1 | 3 - 4 | 326 | 88 | |
GenBank mRNA | BC102369.1 | - | 3 - 4 | 328 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
